Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ntp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307237.2 | NP_001294166.1 | 4 - 5 | 576 | 109 | |
NM_001307238.2 | NP_001294167.1 | 4 - 5 | 727 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |