Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028199.5 | NP_001023370.1 | 13 - 14 | 2258 | 80 | |
NM_001028200.6 | NP_001023371.1 | 15 - 16 | 4215 | 80 | ||
NM_001047522.4 | NP_001040987.1 | 13 - 14 | 2001 | 80 | ||
NM_069133.5 | NP_501534.2 | 12 - 13 | 1878 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |