Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129317.5 | NP_001122789.1 | 2 - 3 | 601 | 68 | |
NM_001372918.2 | NP_001359633.1 | 15 | 1584 | 68 | ||
NM_001380443.1 | NP_001366997.1 | 9 - 10 | 991 | 68 | ||
NM_001383186.1 | 12 - 13 | 1351 | 68 | |||
GenBank mRNA | AF180735.1 | - | 11 - 12 | 1253 | 68 | |
AF233263.1 | - | 11 - 12 | 1193 | 68 | ||
AF233267.1 | - | 9 - 10 | 991 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |