Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fmo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069567.7 | NP_501968.2 | 2 - 3 | 291 | 89 | |
GenBank mRNA | AJ582070.1 | - | 1 - 2 | 180 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |