Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069710.6 | NP_502111.3 | 27 - 28 | 4663 | 96 | |
GenBank mRNA | AY726541.1 | - | 27 - 28 | 4593 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |