Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hum-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028377.5 | NP_001023548.2 | 11 - 12 | 2775 | 82 | |
NM_001307645.3 | NP_001294574.1 | 11 - 12 | 2775 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |