Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pig-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028249.6 | NP_001023420.2 | 5 - 6 | 835 | 60 | |
NM_001028250.5 | NP_001023421.2 | 5 - 6 | 835 | 60 | ||
NM_001307424.2 | NP_001294353.1 | 5 - 6 | 836 | 60 | ||
NM_001307425.1 | NP_001294354.1 | 5 - 6 | 835 | 60 | ||
NM_001307428.3 | NP_001294357.1 | 1 - 2 | 103 | 60 | ||
NM_001307429.3 | NP_001294358.1 | 1 - 2 | 103 | 60 | ||
NM_001380011.1 | NP_001368238.1 | 1 - 2 | 103 | 60 | ||
NM_001380012.1 | NP_001368239.1 | 1 - 2 | 103 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |