Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067742.7 | NP_500143.1 | 2 - 3 | 681 | 74 | |
GenBank mRNA | AF353302.1 | - | 2 - 3 | 655 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |