Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zag-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068023.7 | NP_500424.3 | 3 - 4 | 814 | 65 | |
GenBank mRNA | AY224511.1 | - | 3 - 4 | 819 | 65 | |
AY289599.1 | - | 3 - 4 | 826 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |