Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hlh-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028240.6 | NP_001023411.1 | 1 - 2 | 222 | 80 | |
NM_068060.5 | NP_500461.1 | 1 - 2 | 222 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |