Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068483.5 | NP_500884.2 | 1 - 2 | 120 | 71 | |
GenBank mRNA | AJ871144.2 | - | 1 - 2 | 119 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |