Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069851.4 | NP_502252.3 | 12 - 13 | 2211 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |