Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fat-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268666.3 | NP_001255595.1 | 2 - 3 | 167 | 83 | |
GenBank mRNA | AF260244.1 | - | 1 - 2 | 114 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |