Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aly-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083228.5 | NP_001076697.1 | 1 - 2 | 235 | 101 | |
NR_003454.1 | 1 - 2 | 210 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |