Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047571.4 | NP_001041036.2 | 4 - 5 | 747 | 140 | |
NM_001047572.4 | NP_001041037.1 | 4 - 5 | 504 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |