Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268846.3 | NP_001255775.1 | 6 - 7 | 767 | 75 | |
NM_001268847.4 | NP_001255776.1 | 5 - 6 | 616 | 75 | ||
NM_001307070.3 | NP_001293999.1 | 5 - 6 | 589 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |