Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snf-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070676.4 | NP_503077.2 | 12 - 13 | 2086 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |