Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srab-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071560.3 | NP_503961.2 | 4 - 5 | 841 | 215 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |