Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-35B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071694.1 | NP_504095.1 | 4 - 5 | 1366 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |