Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071892.9 | NP_504293.2 | 2 - 3 | 324 | 79 | |
NM_171452.2 | NP_741531.1 | 2 - 3 | 351 | 79 | ||
NM_171927.3 | NP_741532.1 | 2 - 3 | 343 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |