Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129504.4 | NP_001122976.1 | 4 - 5 | 609 | 81 | |
NM_071946.9 | NP_504347.2 | 5 - 6 | 678 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |