Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cft-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072062.5 | NP_504463.1 | 13 - 14 | 2395 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |