Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028766.4 | NP_001023937.1 | 5 - 6 | 551 | 113 | |
NM_001028767.7 | NP_001023938.1 | 4 - 5 | 449 | 113 | ||
NM_001383322.1 | 4 - 5 | 498 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |