Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028881.4 | NP_001024052.1 | 2 - 3 | 153 | 92 | |
NM_001028882.5 | NP_001024053.2 | 2 - 3 | 153 | 92 | ||
NM_001307743.4 | NP_001294672.1 | 1 - 2 | 101 | 92 | ||
GenBank mRNA | AF166169.1 | - | 1 - 2 | 101 | 92 | |
AF166170.1 | - | 1 - 2 | 101 | 92 | ||
AF261891.1 | - | 2 - 3 | 169 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |