Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grd-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307786.2 | NP_001294715.1 | 4 - 5 | 489 | 104 | |
NM_001368461.2 | NP_001355526.1 | 4 - 5 | 611 | 104 | ||
NM_001368462.3 | NP_001355524.1 | 4 - 5 | 611 | 104 | ||
NM_001368463.3 | NP_001355525.1 | 5 | 627 | 104 | ||
NM_001368464.3 | NP_001355528.1 | 4 | 438 | 104 | ||
NM_001368465.2 | NP_001355527.1 | 4 - 5 | 489 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |