Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129519.3 | NP_001122991.1 | 1 - 2 | 390 | 73 | |
NM_001276937.3 | NP_001263866.1 | 1 - 2 | 389 | 73 | ||
NM_001373121.3 | NP_001360176.1 | 1 - 2 | 396 | 73 | ||
NM_001383414.1 | 1 - 2 | 390 | 73 | |||
NM_073584.6 | NP_505985.1 | 1 - 2 | 390 | 73 | ||
GenBank mRNA | AF016852.1 | - | 1 - 2 | 412 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |