Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cky-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129377.5 | NP_001122849.1 | 2 - 3 | 797 | 129 | |
GenBank mRNA | AB096001.1 | - | 2 - 3 | 794 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |