Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyb-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028941.4 | NP_001024112.2 | 3 - 4 | 580 | 66 | |
NM_074424.9 | NP_506825.4 | 4 - 5 | 2094 | 66 | ||
GenBank mRNA | U21282.1 | - | 3 - 4 | 583 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |