Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srw-56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028718.1 | NP_001023889.1 | 2 - 3 | 465 | 85 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |