Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074665.3 | NP_507066.1 | 1 - 2 | 974 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |