Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074717.5 | NP_507118.1 | 5 - 6 | 1184 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |