Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075468.6 | NP_507869.1 | 5 - 6 | 1490 | 73 | |
GenBank mRNA | Z38016.1 | - | 1512 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |