Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd-70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269992.2 | NP_001256921.1 | 3 - 4 | 527 | 74 | |
NR_070284.1 | 3 - 4 | 427 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |