Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lsy-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029525.4 | NP_001024696.1 | 4 - 5 | 569 | 73 | |
NM_001029526.4 | NP_001024697.1 | 3 - 4 | 401 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |