Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075992.1 | NP_508393.1 | 3 - 4 | 464 | 94 | |
GenBank mRNA | AY008128.1 | - | 3 - 4 | 464 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |