Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: peb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029739.6 | NP_001024910.1 | 2 - 3 | 222 | 130 | |
NM_001377612.2 | NP_001364564.1 | 3 - 4 | 267 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |