Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fax-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277024.2 | NP_001263953.1 | 3 - 4 | 402 | 81 | |
NM_076146.4 | NP_508547.1 | 3 - 4 | 374 | 81 | ||
GenBank mRNA | AF176087.1 | - | 3 - 4 | 374 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |