Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029222.4 | NP_001024393.1 | 2 - 3 | 200 | 149 | |
NM_001029223.6 | NP_001024394.1 | 1 - 2 | 103 | 149 | ||
GenBank mRNA | D86740.1 | - | 222 | 149 | ||
D86741.1 | - | 222 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |