Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076396.7 | NP_508797.3 | 1 - 2 | 169 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |