Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mig-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029489.6 | NP_001024660.1 | 4 - 5 | 495 | 98 | |
NM_001029490.5 | NP_001024661.1 | 2 - 3 | 381 | 98 | ||
GenBank mRNA | AF150958.1 | - | 3 - 4 | 399 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |