Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lfi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375027.2 | NP_001362033.1 | 12 - 13 | 5123 | 118 | |
NM_001375029.2 | NP_001362032.1 | 11 - 12 | 4916 | 118 | ||
NM_001375030.2 | NP_001362152.1 | 11 - 12 | 5074 | 118 | ||
NM_001375031.2 | NP_001362153.1 | 10 - 11 | 4824 | 118 | ||
NM_001383570.1 | 12 - 13 | 5427 | 118 | |||
NM_076447.6 | NP_508848.2 | 13 - 14 | 5461 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |