Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abts-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029655.6 | NP_001024826.2 | 19 - 20 | 2815 | 128 | |
NM_001129697.5 | NP_001123169.1 | 18 - 19 | 2755 | 128 | ||
NM_001373337.3 | NP_001360088.1 | 18 - 19 | 2739 | 128 | ||
GenBank mRNA | AY887906.1 | - | 19 - 20 | 2837 | 128 | |
AY954522.1 | - | 18 - 19 | 2777 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |