Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pes-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076766.9 | NP_509167.2 | 3 - 4 | 723 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |