Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mnm-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076855.4 | NP_509256.2 | 5 - 6 | 697 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |