Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mgl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129721.4 | NP_001123193.2 | 18 - 19 | 2661 | 74 | |
NM_001313458.3 | NP_001300387.1 | 19 - 20 | 2703 | 74 | ||
NM_001313459.3 | NP_001300388.1 | 15 - 16 | 2436 | 74 | ||
NM_077273.4 | NP_509674.2 | 19 - 20 | 2920 | 74 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |