Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pxn-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077433.5 | NP_509834.1 | 6 - 7 | 810 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |