Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: odr-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077688.4 | NP_510089.1 | 4 - 5 | 1079 | 94 | |
GenBank mRNA | U16708.1 | - | 4 - 5 | 1086 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |