Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307870.3 | NP_001294799.1 | 4 - 5 | 712 | 78 | |
NM_001307871.5 | NP_001294800.1 | 3 - 4 | 482 | 78 | ||
NM_001373568.1 | NP_001360629.1 | 5 - 6 | 778 | 78 | ||
NM_077965.5 | NP_510366.1 | 4 - 5 | 589 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |