Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hrg-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028013.2 | NP_001023184.1 | 3 - 4 | 434 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |