Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268567.3 | NP_001255496.1 | 2 - 3 | 465 | 109 | |
NM_001268568.3 | NP_001255497.1 | 1 - 2 | 131 | 109 | ||
NR_131602.1 | 2 - 3 | 236 | 109 | |||
NR_131603.1 | 2 - 3 | 187 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |